Thermo Scientific™

CellSensor™ AP-1-bla HEK293T Cell Line

Catalog number: K1658
Have Questions?
Thermo Scientific™

CellSensor™ AP-1-bla HEK293T Cell Line

Catalog number: K1658
Have Questions?
Catalog Number
K1658
Unit Size
1 vial
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K16581 vial
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The CellSensor® AP-1-bla HEK 293T Cell Line contains a beta-lactamase reporter gene under control of the AP-1 response element stably integrated into HEK 293T cells. The cell line was created through sorting by FACS of cells responsive to stimulation of the AP-1 pathway with phorbol 12-myristate 13-acetate (PMA). The cell line was validated for DMSO tolerance, incubation time with stimulant, and substrate loading conditions. The AP-1-bla HEK 293T cell line responds to agonist treatment as expected from literature and can be adapted for high throughput screening for agonists or antagonists of the AP-1 pathway with compound libraries. Candidate drugs can also be tested for dose response against this cell line. Academic and non-profit customers, please inquire for special pricing.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Entry
Cell-based beta lactamase reporter gene
Cell State
Dividing Cells
Product Line
CellSensor™
Target Entry
EGFR, ERK1/2, p38, MEK, MAPK pathway, c-MET, JNK, RON, RAF
System Type
CellSensor™
Druggable Target
Signaling Pathway

Contents & Storage

CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text