Thermo Scientific™

LanthaScreen™ c-Jun (1-79) HeLa Cell Line

Catalog number: K1680
Have Questions?
Thermo Scientific™

LanthaScreen™ c-Jun (1-79) HeLa Cell Line

Catalog number: K1680
Have Questions?
Catalog Number
K1680
Unit Size
1 vial
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K16801 vial
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The LanthaScreen®c-Jun (1-79) HeLa cell line contains a fusion protein consisting of GFP and a fragment encoding for AA 1-79 of c-Jun under the control of a CMV promoter stably transfected into HeLa cell lines. HeLa is a cervical cancer cell line which shows inducible activation of JNK by a number of different ligands, such as TNF-a and EGF, which leads to the transient phosphorylation of GFP-c-Jun (1-79). This LanthaScreen®cell line therefore allows the analysis of JNK activity using GFP-c-Jun phosphorylation as readout. The GFP-c-Jun (1-79) construct was transfected into HeLa cells using Lipofectamine™2000, followed by selection with Blasticidin. This cell line is a clonal population isolated by flow cytometry using GFP fluorescence as sorting marker. This cell line is validated for EC50 and Z' under optimized conditions using TNF-a as ligand for JNK mediated GFP-c-Jun (1-79) phosphorylation. This cell lines has also been tested for assay performance under variable experimental conditions, including cell plating density, stimulation time, DMSO tolerance and assay development time. Additional information using alternate stimuli is also available. Academic and non-profit customers, please inquire for special pricing.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Entry
Cell-based terbium phospho-specific activity (stable)
Cell State
Dividing Cells
Product Line
LanthaScreen™
Target Entry
c-Jun
System Type
LanthaScreen™
Druggable Target
Signaling Pathway

Contents & Storage

LanthaScreen™ Cellular assays are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text