Thermo Scientific™

GeneBLAzer™ RXRα-UAS-bla HEK 293T Cells

Catalog number: K1697
Have Questions?
Thermo Scientific™

GeneBLAzer™ RXRα-UAS-bla HEK 293T Cells

Catalog number: K1697
Have Questions?
Catalog Number
K1697
Unit Size
2 x 106 cells
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K16972 x 106 cells
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

RXR alpha-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human retinoid X receptor-alpha (RXR alpha) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UASbla HEK 293T cell line. GeneBLAzer™UAS-bla HEK 293T cells stably express a betalactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-RXR alpha (LBD) fusion protein, the protein binds to the UAS, resulting in expression of alpha-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. RXR alpha-UAS-bla HEK 293T cells are functionally validated for Z' and EC50 concentrations of 9-cis-retinoic acid. In addition, RXR alpha-UAS-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Alternate Agonists
Dexamethasone, Progesterone, Cortisol
Antagonists
Spironolactone
Assay Entry
Cell-based beta lactamase reporter gene
Cell State
Dividing Cells
Detection Method
Fluorescence
Species
Human
Druggable Target
Nuclear Receptors
Primary Agonist
Aldosterone
Product Line
GeneBLAzer™
Product Type
HEK 293T Cells
Quantity
2 x 106 Cells
Reporter Gene
BLA (Beta-Lactamase)
Target Entry
RXRA, RXR alpha, NR2B1

Contents & Storage

RXR alpha-UAS-bla HEK 293T cells (K1261)

Shipping: Dry Ice
Storage: -80°C (liquid nitrogen)

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text