Tango™ GPCR Enabling Model

Catalog number: K1820
Have Questions?

Tango™ GPCR Enabling Model

Catalog number: K1820
Have Questions?
Catalog Number
K1820
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K18201 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Tango™ GPCR Enabling Model provides in an easy to use system the same reagents Invitrogen uses to make the family of Tango™ GPCR cell lines:

1. The Tango™ Arrestin2-UAS-bla U20S parental cell line that has the beta-lactamase (bla) reporter gene under the control of a UAS response element stably integrated as well as the beta-arrestin⁄TEV protease fusion protein constitutively expressed.

2.The Tango™ pDest-Gal4VP16-M vector which is used for insertion of your GPCR of interest for in the Tango™ Arrestin2-UAS-bla U2OS parental cell line.

When the Tango™ pDest-Gal4VP16-M vector containing a GPCR of interest is integrated into the parental cell line, the chimeric receptor-Gal4VP16 protein is expressed. Upon ligand binding and receptor activation the protease tagged arrestin is recruited to the receptor. The UAS response element is cleaved from the GPCR and translocates to the nucleus and drives the beta-lactamase reporter gene expression. Activation of the reporter gene provides a quantifiable measurement of the degree of interaction between the target receptor and the protease-tagged arrestin partner and is unaffected by other signaling pathways in the cell, providing an exquisitely specific readout of target receptor activation.

Invitrogen's Tango™ GPCR Assay technology combines the benefits of the Tango™ assay platform with the highly accurate, sensitive and live-cell beta-lactamase reporter system. The Tango™ assay platform is based upon a ligand binding to a G Protein-Coupled Receptor (GPCRs) that triggers desensitization, a process mediated by the recruitment of intracellular arrestin proteins to the activated receptor. As a result, the ligand-induced activation of GPCRs may be assayed by monitoring the interaction of arrestin with the test GPCR. A major advantage of this approach is that it does not depend on the G protein signaling specificity of the target receptor.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell State
Dividing Cells
Detection Method
Fluorescent
For Use With (Application)
FRET
Human Protein Class
G-Protein Coupled Receptor (GPCR)
Product Line
GeneBLAzer™, Tango™
Readout
End Point
Cell Line
U2OS
System Type
Tango™
Druggable Target
GPCR

Contents & Storage

Contents:
K1820 Tango™ GPCR Enabling Model
(LiveBLAzer™-FRET B⁄G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
• Tango™ Arrestin2-UAS-bla U2OS cells
• Tango™ pDest Gal4VP16-M Vector
• Tango™ Enabling Model Assay Protocol
• Certificate of Analysis

Storage:
The Tango™ Arrestin2-UAS-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. The Tango™ pDest Gal4VP16-M vector is shipped on dry ice and should be stored at -20 immediately upon receipt.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text