Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

FastDigest Value Pack

Catalog number: K1991
Have Questions?
Thermo Scientific™

FastDigest Value Pack

Catalog number: K1991
Have Questions?
Catalog Number
K1991
Unit Size
20 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K199120 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific FastDigest Value Pack contains the 13 most commonly used FastDigest enzymes. The selection of enzymes will support a wide range of molecular biology applications such as vector and insert preparation for cloning, fast clone analysis and digestion of PCR products. Each enzyme in the pack allows for 20 standard restriction digest reactions.

Thermo Scientific FastDigest enzymes are an advanced line of restriction enzymes that are all 100% active in a single buffer. The universal FastDigest and FastDigest Green buffer allows single-, double- or multiple DNA digestion within 5–15 minutes eliminating any need for buffer change or subsequent DNA clean-up steps. Thermo Scientific DNA modifying enzymes have 100% activity in this buffer as well. For additional convenience the FastDigest Green Buffer includes a density reagent and two tracking dyes which allows for direct loading of digestion reaction products on gels.

Highlights

• 100% activity of all FastDigest enzymes in a single buffer
• Complete digestion in 5–15 minutes and direct loading on gels
• 100% buffer compatibility with downstream applications
• 176 FastDigest enzymes available
• No star activity

Note

The FastDigest Green Buffer offers the same high performance in DNA digestion and downstream applications as the colorless FastDigest Buffer. For applications that require product analysis by fluorescence excitation (e.g. concentration measurements in UV light) the colorless FastDigest Buffer is recommended.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Enzyme
EcoR I, Bgl II, Nde I, Sal I, Xba I, Kpn I, Eco32I, Xho I, Not I, BamH I, Hind III, Sma I, Pst I
Compatible Buffer
10X FastDigest Buffer/FastDigest Green Buffer
Type IIS RE
No
Quantity
13 x 20 μL (13 x 20 Reactions)
Product Type
Value Pack
Research Category
Traditional Cloning
Product Line
FastDigest™
Overhang
Varied based on enzyme
Inactivation Temperature
Varies based on enzyme

Contents & Storage

• FastDigest enzyme (13 enzymes at 20 μL each)
• 10X FastDigest Buffer (2 x 1.0 mL)
• 10X FastDigest Green Buffer (2 x 1.0 mL)

Store at –20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text