PureLink™ Quick Plasmid Miniprep Kit
For better long-term performance store purification columns at 2°C to 8°C.
PureLink™ Quick Plasmid Miniprep Kit
Invitrogen™

PureLink™ Quick Plasmid Miniprep Kit

The PureLink Quick Plasmid Miniprep Kit is designed for rapid isolation of sequencing-grade plasmid DNA from E.coli in about 30 minutes, using silica membrane columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
K21001050 Preps
K210011250 Preps
Catalog number K210010
Price (USD)
108.65
Online Exclusive
112.00
Save 3.35 (3%)
Each
Add to cart
No. of Reactions:
50 Preps
Price (USD)
108.65
Online Exclusive
112.00
Save 3.35 (3%)
Each
Add to cart

The PureLink™ Quick Plasmid Miniprep Kit is designed for rapid isolation of sequencing-grade plasmid DNA from E.coli in 30-45 minutes, using silica membrane columns.

Features of the PureLink™ Quick Plasmid Miniprep Kit:

  • Fast—ready-to-use plasmid DNA typically in 30-45 minutes
  • Flexible—formats work with vacuum and centrifugation platforms

Reliable and cost-effective isolation of plasmid DNA

The PureLink™ Quick Plasmid Miniprep Kit allows isolation of high-quality plasmid DNA from 1–5 mL cultures in 30-45 minutes, using the PureLink™ spin columns. These columns use a unique silica membrane to achieve high yields of up to 40 μg of sequencing-grade plasmid DNA. Cells are lysed using an alkaline/SDS procedure and the lysate is applied to a silica membrane column that selectively binds plasmid DNA. Contaminants are removed with Wash Buffer, and the plasmid DNA is eluted in TE Buffer. The eluted DNA is suitable for all routine downstream applications.

Compatible with flexible formats

The PureLink™ Quick Plasmid Miniprep Kit can be used with a centrifuge or a vacuum manifold for purifying plasmid DNA. Typically, DNA isolated using the PureLink™ Quick Plasmid Miniprep Kit has an A260/A280 of >1.80 when samples are diluted in Tris-HCl (pH 7.5), indicating that the DNA is reasonably free of proteins that could interfere with downstream applications.

Isolated DNA can be used for a variety of downstream applications

Plasmid DNA purified using PureLink™ Quick Plasmid Miniprep Kit is suitable for all routine downstream applications, including bacterial and mammalian cell transformations, DNA sequencing, restriction enzyme digestion, cloning, and PCR.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeSpin Column
Concentration20 mg/mL
Final Product TypePlasmid DNA
For Use With (Application)Next-Generation Sequencing, Cloning, Sequencing, Nucleic Acid Labeling, PCR, Transformation
High-throughput CompatibilityNot High-throughput Compatible (Manual)
No. of Reactions50 Preps
Prep Scale< 100 μg (Small-Scale) Plasmid DNA
Product LinePureLink™
Product TypeQuick Plasmid MiniPrep Kit
Quantity50 Preps
Sample TypeBacterial Culture
Shipping ConditionRoom Temperature
TargetPlasmid DNA
Test Time30 to 45 min.
FormatSpin Column
Isolation TechnologySilica Spin Column
Unit SizeEach
Contents & Storage
• Resuspension buffer
• RNase A (20 mg/mL)
• Lysis buffer
• Precipitation buffer
• Wash buffers
• TE buffer
• Spin columns in wash tubes
• Elution tubes

Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link