PureLink™ Nucleic Acid Purification Rack
PureLink™ Nucleic Acid Purification Rack
Invitrogen™
PureLink™ Nucleic Acid Purification Rack
Convenient large capacity fits multiple-sized column• The compact nesting feature is convenient for storageThe PureLink™ Nucleic Acid Purification Rack isRead more
Have Questions?
Catalog NumberQuantity
K2100131 Ea.
Catalog number K210013
Price (USD)
432.65
Online Exclusive
454.00
Save 21.35 (5%)
Each
Add to cart
Quantity:
1 Ea.
Price (USD)
432.65
Online Exclusive
454.00
Save 21.35 (5%)
Each
Add to cart
Convenient large capacity fits multiple-sized column
• The compact nesting feature is convenient for storage
The PureLink™ Nucleic Acid Purification Rack is designed for use with columns in the PureLink™ HiPure Plasmid Purification Systems. The rack includes both a Collection Tube Rack for holding various sized recovery tubes (Figure 1) and a large-capacity Waste Reservoir. One rack holds as many as 12 miniprep columns, 8 midiprep columns, and 4 maxiprep columns. When you're finished, the Waste Reservoir nests into to the Collection Tube Rack for compact storage.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LinePureLink™
Quantity1 Ea.
Shipping ConditionRoom Temperature
FormatRack
Product TypeNucleic Acid Purification Rack
Unit SizeEach
Contents & Storage
The PureLink™ Nucleic Acid Purification Rack includes:
• Column Holder Rack (for processing 12 miniprep, 8 midiprep, and 4 maxiprep columns)
• Collection Tube Rack (capable of accommodating various types and sizes of recovery tubes)
• Large Capacity Waste Tray (for collecting waste)
Frequently asked questions (FAQs)
How do I eliminate the RNA contamination I get in my minipreps?
Low copy number plasmids may be contaminated with RNA due to the large number of cells being processed. Increase the concentration of RNAse in the buffers 4 fold.
Share catalog number, name or link
1x1 image pixel for data collection