Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

TransformAid Bacterial Transformation Kit

Catalog number: K2711
Have Questions?
Thermo Scientific™

TransformAid Bacterial Transformation Kit

Catalog number: K2711
Have Questions?
Catalog Number
K2711
Unit Size
40 transformations
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K271140 transformations
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific TransformAid Bacterial Transformation Kit uses a novel method for rapid preparation of chemically-competent E. coli cells from overnight bacterial cultures or bacterial colonies. The key component of this system is the unique T-solution, which produces competent cells in a few easy steps. The quick and convenient procedure guarantees transformation efficiencies of more than 107 transformants per μg of plasmid DNA, ideal for routine cloning experiments. The option to use bacterial colonies for preparation of competent cells adds flexibility to any cloning experiment.

The TransformAid Bacterial Transformation Kit can be used with most E. coli strains commonly used for cloning: JM107, JM109, XL1-Blue, SURE, TOPP2, W3110, NM527, AD494, CJ236, GM2163, C600, BL21, M15, NM522, HB101, ER2267.

Highlights

• Efficient—more than 107 transformants per μg of plasmid DNA
• Practical—E. coli competent cells can be prepared from an overnight culture or from bacterial colonies
• Fast—less than 1 hour from the overnight bacterial culture to cell plating. Starting from bacterial colonies, the procedure requires only 2.5 hours
• Easy—all procedures are performed on ice on your bench. Brief centrifugations are carried out in regular microcentrifuges

Applications

• Routine cloning experiments
• Blunt-end cloning
• TA cloning

Includes

• C-Medium
• T-Solution (A)
• T-Solution (B)
• Detailed Protocol

Note

Competent cells prepared with TransformAid Bacterial Transformation Kit are suitable for direct-use only. Freezing down and storage at -70°C is not recommended.

Related Products
TransformAid Bacterial Transformation Kit
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
E. coli
Format
Kit
Species
E. coli
Efficiency
≥1 x 107;
For Use With (Application)
Cloning
No. of Reactions
40 Reactions
Product Line
TransformAid
Product Type
TransformAid Bacterial Transformation Kit
Quantity
40 Transformations

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text