Invitrogen™

Zero Blunt™ TOPO™ PCR Cloning Kit, with One Shot™ TOP10 Electrocomp™ E. coli

Catalog number: K286020
Have Questions?
Invitrogen™

Zero Blunt™ TOPO™ PCR Cloning Kit, with One Shot™ TOP10 Electrocomp™ E. coli

Catalog number: K286020
Have Questions?
Catalog Number
K286020
Unit Size
25 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K28602025 reactions
-
K28604050 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Zero Blunt™ TOPO™ PCR Cloning Kits offers the fastest and easiest method for high-efficiency (cloning of blunt-end PCR products amplified with proofreading thermostable polymerases. The kits include linearized and topoisomerase I-activated pCR™-Blunt II-TOPO™ Vector for 5-minute benchtop ligations without ligase (1). The pCR™-Blunt II-TOPO™ Vector also features:

ccdB gene for positive selection (2)
EcoR I sites flanking the PCR product insertion site for easy excision of inserts
• Kanamycin and Zeocin™ resistance genes for your choice of selection in E. coli
• SP6 promoter/primer site for in vitro RNA transcription and sequencing
• M13 forward and reverse primer sites for sequencing or PCR screening

Selected Zero Blunt™ TOPO™ PCR Cloning Kits are available in combo kits combined with a PureLink™ Quick Plasmid Miniprep Kit (50 preps) for fast plasmid purification of your TOPO™-cloned inserts for downstream analysis.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Bacterial or Yeast Strain
TOP10
Cloning Method
Blunt TOPO™
Format
Kit
Product Line
One Shot™
Product Type
PCR Cloning Kit
Promoter
SP6
Quantity
25 Reactions
Vector
Blunt DNA Cloning Vectors
Unit Size
25 reactions

Contents & Storage

Box 1:
• Linearized and topoisomerase I-activated pCR™Blunt II-TOPO™ vector
• Salt solution
• dNTPs
• Control template
• M13 forward and reverse primers
• Sterile water

Store at -5 to -30°C.
All reagents are stable for 6 months when properly stored.

Box 2:
• One Shot™ Chemically Competent or Electrocomp™ E. coli
• S.O.C. medium
• Supercoiled pUC19 control plasmid

Store at -68 to -85°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text