Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:Informational:Need Ethanol or Isopropanol for your nucleic acid isolation? Visit our Molecular Biology Grade Solvents page.
Invitrogen™

PureLink™ PCR Purification Kit

Catalog number: K310001
Have Questions?
Invitrogen™

PureLink™ PCR Purification Kit

Catalog number: K310001
Have Questions?
Catalog Number
K310001
Unit Size
50 preps
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
250 preps
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K31000150 preps
-
K310002250 preps
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

The PureLink PCR Purification Kit provides a more convenient, rapid, and high-yielding purification process compared to other companies' kits. After PCR amplification, purification of the PCR amplicons is required for many common downstream applications, including enzyme digestion, ligation, sequencing, and labeling experiments, to prevent enzymes (polymerase, restriction enzyme) and reaction components (dNTPs, primers, buffers, salts) from carrying over, contaminating, and negatively influencing the downstream experiments.

Advantages of the PureLink PCR Purification Kit include:
• 4X binding capacity compared to Company Q; 40 μg for the PureLink kit versus 10 μg from other companies' kits
• No need for tedious sample pH adjustments necessary with other kits
• Convenient and rapid protocol, completed in less than 10 minutes
• Optional size-selection buffers included for selectively purifying 300–600 bp fragments or removing fragments <300 bp

The PureLink PCR Purification Kit is based on the selective binding of dsDNA to silica-based membrane in the presence of chaotropic salts. Simply mix the PCR product with Binding Buffer and apply to the PureLink Spin Column. The dsDNA binds to the silica-based membrane in the column. Remove impurities by thorough washing with Wash Buffer. To purify the DNA, elute the dsDNA in low-salt Elution Buffer or water.The purified PCR product is suitable for automated fluorescent DNA sequencing, restriction enzyme digestion, and cloning.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Kit
High-throughput Compatibility
High-throughput Compatible
Product Line
PureLink™
Product Type
PCR Purification Kit
Quantity
50 Preps
Run Time
10 min.
Shipping Condition
Room Temperature
Target
DNA from Enzymatic Reactions (e.g. PCR)
For Use With (Application)
Next-Generation Sequencing (NGS), Cloning
Isolation Technology
Silica Spin Column
Sample Type
PCR Products

Contents & Storage

• 15 mL Binding Buffer (B2)
• 23 mL Binding Buffer High-Cutoff (B3)
• 16 mL Wash Buffer (W1)
• 15 mL Elution Buffer (E1)
• 50 PureLink™ PCR Spin Columns with Collection Tubes
• 50 PureLink™ Elution Tubes (1.7 mL)

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text