Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:Informational:Need Ethanol or Isopropanol for your nucleic acid isolation? Visit our Molecular Biology Grade Solvents page.
Invitrogen™

PureLink™ PCR Micro Kit

Catalog number: K310050
Have Questions?
Invitrogen™

PureLink™ PCR Micro Kit

Catalog number: K310050
Have Questions?
Catalog Number
K310050
Unit Size
50 preps
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
250 preps
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K31005050 preps
-
K310250250 preps
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

PureLink PCR Micro Kits are ideal for purifying and concentrating DNA from low-yielding PCR or restriction digestion reactions. These highly purified DNA fragments and amplicons provide excellent results in downstream applications such as DNA sequencing, restriction digestion, cloning, PCR, and labeling. PureLink PCR Micro Kits provide a simple and rapid method for purifying and concentrating DNA fragments ranging in size from 100 bp-12 kb.

The unique micro-nozzle design allows up to 95% recovery of DNA fragments with as little as a 5 μL elution volume. With less than a 10% coefficient of variance in the elution volume you can be sure that no ethanol is carried over from the wash steps giving you the best chance of success in downstream applications. Removal of primers, dNTPs, enzymes, and salts exceeds 98.5%. This kit can also efficiently remove unincorporated dye-labeled nucleotides from a cDNA labeling reaction.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Spin Column
Elution Volume Max
20 μL
Elution Volume Min
5 μL
High-throughput Compatibility
Not High-throughput Compatible (Manual)
Product Line
PureLink™
Product Type
PCR Micro Kit
Quantity
50 Preps
Run Time
10 min.
Separation Range
100 bp to 12 kb
Shipping Condition
Room Temperature
Starting Material Volume
125 μL
Target
DNA from Enzymatic Reactions (e.g. PCR)
For Use With (Application)
Cloning, Standard PCR
Isolation Technology
Silica Spin Column
No. of Reactions
50 Preps
Sample Type
cDNA Labeling Reactions

Contents & Storage

Kit includes columns, binding and wash buffers, recovery tubes, and the manual. Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text