Invitrogen™

TOPO™ TA Cloning™ Kit, Dual Promoter, with One Shot™ MAX Efficiency™ DH5α-T1R E. coli

Catalog number: K462040
Have Questions?
Invitrogen™

TOPO™ TA Cloning™ Kit, Dual Promoter, with One Shot™ MAX Efficiency™ DH5α-T1R E. coli

Catalog number: K462040
Have Questions?
Catalog Number
K462040
Unit Size
50 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
25 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K46204050 reactions
-
K46200125 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
TOPO™ TA Cloning™ Kits are designed for cloning PCR products directly from a PCR reaction in just 5 minutes (1). They use a pCR™-TOPO™ Vector with covalently bound topoisomerase I for fast cloning and recombinants. pCR™-TOPO™ Vectors include:

• 3´-T overhangs for direct ligation of Taq-amplified PCR products
• Choice of T7 (pCR™2.1-TOPO™) or T7 and SP6 (pCR™II-TOPO™) promoters for in vitro RNA transcription and sequencing. Each vector also contains M13 forward and reverse primer sites for sequencing.
EcoR I sites flanking the PCR product insertion site for easy excision of inserts
• Kanamycin and ampicillin resistance genes for your choice of selection in E. coli
• Easy blue/white colony screening for selection of recombinants

TOPO™ TA Cloning™ Kits are also available in combo kits combined with a PureLink™ Quick Plasmid Miniprep Kit (50 preps) for fast plasmid purification of your TOPO™-cloned inserts for downstream analysis.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Bacterial or Yeast Strain
DH5α
Cloning Method
TOPO™-TA
Format
Kit
Product Line
One Shot™
Product Type
Cloning Kit
Promoter
T7, SP6
Quantity
50 Reactions
Vector
TOPO-TA Cloning Vectors

Contents & Storage

Box 1:
• Topoisomerase I-activated pCR™II-TOPO™ vector
• PCR buffer
• Salt solution
• dNTPs
• Control template
• M13 forward and reverse primers
• Control PCR primers
• Sterile water

Store at -5 to -30°C.
All reagents are stable for 6 months when properly stored.

Box 2:
• One Shot™ Chemically Competent or Electrocomp™ E. coli
• S.O.C. medium
• Supercoiled pUC19 control plasmid.

Store at -68 to -85°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text