Invitrogen™
Expressway™ Mini Cell-Free Expression System
Catalog number: K990100
Have Questions?
Invitrogen™
Expressway™ Mini Cell-Free Expression System
Catalog number: K990100
Have Questions?
Catalog Number
K990100
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K9901001 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Expressway™ Mini Cell-Free Expression System utilizes an efficient coupled transcription and translation reaction to produce active recombinant protein. This system eliminates the time-consuming steps of cell-based protein production including transformation, cell culture, and expression optimization. In just two hours, you can produce protein suitable for an array of downstream applications and functional studies. The Expressway™ Mini Cell-Free Expression System offers flexibility in experimental design, allowing you to
• Synthesize proteins from both circular plasmid and linear templates (e.g. PCR products)
• Perform high-throughput synthesis of proteins
• Express genes toxic to in vivo systems
How it works:
The Expressway™ Mini Cell-Free Expression System provides all the components you need for optimal cell-free protein production. The kit includes an E. coli extract containing the cellular machinery required to drive transcription and translation. An IVPS (in vitro protein synthesis) reaction buffer and an amino acid module are also included in the kit to provide the required amino acids and an ATP-regenerating system. Simply mix the reaction buffer, amino acid mix, methionine (labeled or unlabeled), T7 Enzyme Mix, and your DNA template (containing a T7 promoter) with the E. coli extract. As the DNA template is transcribed, the 5’ end of the mRNA is bound by ribosomes and undergoes translation as the 3’ end of the template is still being transcribed (Figure 1).
Functional protein made easy:
The Expressway™ Mini Cell-Free Expression System has been optimized to produce high yields of full-length, functional protein from E. coli cell lysates (Figure 2). A proprietary feed buffer formulation is provided with the system for increased protein yields.
Contents and Storage:
The Expressway™ Mini Cell-Free Expression System is designed to perform twenty 50-μl reactions or one 1-ml reaction. This system includes the IVPS E.coli Extract, IVPS reaction buffer, feed buffer, 19 amino acid mix, methionine, DNase/RNase-free water, RNase A, T7 Enzyme Mix, 2-ml reaction tubes, and a positive expression control vector. Suggested vectors for use with the Expressway™ Mini Cell-Free Expression System are listed below. Store vectors at -20 °C. Store other kit components at -80 °C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Promoter
T7
Product Type
Cell-Free Expression System
Expressed Protein Type
Water Soluble Protein
Expression System
Cell-Free
Expression Mechanism
Cell-Free Expression
Product Line
Expressway™