Thermo Scientific Chemicals

1,3-Diphenylisobenzofuran, 97%, Thermo Scientific Chemicals

Catalog number: L00101.06
5 g, Each
Thermo Scientific Chemicals

1,3-Diphenylisobenzofuran, 97%, Thermo Scientific Chemicals

Catalog number: L00101.06
5 g, Each
Quantity
Catalog number: L00101.06
also known as L00101-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5471-63-6
IUPAC Name
1,3-diphenyl-2-benzofuran
Molecular Formula
C20H14O
InChI Key
ZKSVYBRJSMBDMV-UHFFFAOYSA-N
SMILES
O1C(=C2C=CC=CC2=C1C1=CC=CC=C1)C1=CC=CC=C1
Form
Crystals or crystalline powder
Melting Point (clear melt)
128-137?C
Assay (GC)
≥96.0%
Appearance (Color)
Yellow to green
Assay (HPLC)
≥96.0%

Description

1,3-Diphenylisobenzofuran is used as a fluorescent probe for detection of superoxide anion radical (O2-) inside the membrane lipid layer by DPBF fluorescence quenching method. It is used as quencher during the photo inactivation of TA-3 mouse mammary carcinoma cells containing hematoporphyrin. 1,3-Diphenylisobenzofuran is used to study the single crystal molecular structure and solution photophysical properties of DPBF.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diphenylisobenzofuran is used as a fluorescent probe for detection of superoxide anion radical (O2-) inside the membrane lipid layer by DPBF fluorescence quenching method. It is used as quencher during the photo inactivation of TA-3 mouse mammary carcinoma cells containing hematoporphyrin. 1,3-Diphenylisobenzofuran is used to study the single crystal molecular structure and solution photophysical properties of DPBF.

Solubility
Soluble in toluene. Insoluble in water.

Notes
Air & Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text