Thermo Scientific Chemicals

1-(Trifluoroacetyl)imidazole, 98+%, Thermo Scientific Chemicals

Catalog number: L00135.09
10 g, Each
Thermo Scientific Chemicals

1-(Trifluoroacetyl)imidazole, 98+%, Thermo Scientific Chemicals

Catalog number: L00135.09
10 g, Each
Quantity
Catalog number: L00135.09
also known as L00135-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1546-79-8
IUPAC Name
2,2,2-trifluoro-1-(1H-imidazol-1-yl)ethan-1-one
Molecular Formula
C5H3F3N2O
InChI Key
SINBGNJPYWNUQI-UHFFFAOYSA-N
SMILES
FC(F)(F)C(=O)N1C=CN=C1
Appearance (Color)
Clear colorless to yellow to pale orange or pink
Refractive Index
1.4225-1.4265 @ 20°C (non-U.S. specification)
Assay (GC)
≥98.0%
Form
Liquid
Comment
Specification differs for U.S. and non-U.S. material where indicated

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text