Thermo Scientific™

N,O-Bis(trimethylsilyl)acetamide, 95%, Thermo Scientific Chemicals

Catalog number: L00183.14
25 g, Each
Thermo Scientific™

N,O-Bis(trimethylsilyl)acetamide, 95%, Thermo Scientific Chemicals

Catalog number: L00183.14
25 g, Each
Quantity
Catalog number: L00183.14
also known as L00183-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10416-59-8
IUPAC Name
trimethylsilyl N-(trimethylsilyl)ethanimidate
Molecular Formula
C8H21NOSi2
InChI Key
SIOVKLKJSOKLIF-UHFFFAOYSA-N
SMILES
CC(O[Si](C)(C)C)=N[Si](C)(C)C
Appearance (Color)
Clear, colorless to pale yellow
Form
Liquid
Assay (GC)
≥94.0%

Description

N,O-Bis(trimethylsilyl)acetamide is used as a regioselective desulfation reagent as well as a preparatory agent for carbohydrate and alcohol trimethylsilyl ethers. It is also used for the derivatization of polar functional groups such as carboxylic acids, phenols, steroids, amines, alcohols, alkaloids and amides. It is employed in the formation of stable trimethylsilyl derivatives. Further, it is used in analytical chemistry for the derivatization of compounds in analysis to increase their volatility and to introduce the trimethylsilyl protecting group in organic synthesis. In addition to this, it is used in the protection of hydroxyl groups in natural products, functional groups in organic intermediates and derivatization reagent for gas chromatography-Mass spectral analysis of phenolic acids in fruits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,O-Bis(trimethylsilyl)acetamide is used as a regioselective desulfation reagent as well as a preparatory agent for carbohydrate and alcohol trimethylsilyl ethers. It is also used for the derivatization of polar functional groups such as carboxylic acids, phenols, steroids, amines, alcohols, alkaloids and amides. It is employed in the formation of stable trimethylsilyl derivatives. Further, it is used in analytical chemistry for the derivatization of compounds in analysis to increase their volatility and to introduce the trimethylsilyl protecting group in organic synthesis. In addition to this, it is used in the protection of hydroxyl groups in natural products, functional groups in organic intermediates and derivatization reagent for gas chromatography-Mass spectral analysis of phenolic acids in fruits.

Solubility
Miscible with many nonpolar and polar aprotic solvents.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, water and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text