N,N-Dimethylformamide diethyl acetal, 95%, Thermo Scientific Chemicals
N,N-Dimethylformamide diethyl acetal, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Dimethylformamide diethyl acetal, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L00205.09
also known as L00205-09
Price (USD)/ Each
34.50
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
34.50
Add to cart
N,N-Dimethylformamide diethyl acetal, 95%, Thermo Scientific Chemicals
Catalog numberL00205.09
Price (USD)/ Each
34.50
-
Add to cart
Chemical Identifiers
CAS1188-33-6
IUPAC Name(diethoxymethyl)dimethylamine
Molecular FormulaC7H17NO2
InChI KeyBWKAYBPLDRWMCJ-UHFFFAOYSA-N
SMILESCCOC(OCC)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless to pale yellow
Formliquid
Assay (GC)>94.0%
Refractive Index1.3960-1.4020
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only