Thermo Scientific Chemicals

1-(Heptafluorobutyryl)imidazole, 97%, Thermo Scientific Chemicals

Catalog number: L00221.06
5 g, Each
Thermo Scientific Chemicals

1-(Heptafluorobutyryl)imidazole, 97%, Thermo Scientific Chemicals

Catalog number: L00221.06
5 g, Each
Quantity
Catalog number: L00221.06
also known as L00221-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
32477-35-3
IUPAC Name
2,2,3,3,4,4,4-heptafluoro-1-(1H-imidazol-1-yl)butan-1-one
Molecular Formula
C7H3F7N2O
InChI Key
MSYHGYDAVLDKCE-UHFFFAOYSA-N
SMILES
FC(F)(F)C(F)(F)C(F)(F)C(=O)N1C=CN=C1
Appearance (Color)
Clear to faintly turbid colorless to pale yellow to pink
Refractive Index
1.3825-1.3875 @20°C
Assay (GC)
≥96.0%
Form
Liquid

Description

1-(Heptafluorobutyryl)imidazole is used in gas chromatography for the determination of various pharmaceutical compounds like retronecine in biological matrixes. It acts as a reagent for derivatize amine-groups. It plays an essential role to reduce GC column degradation due to its non-acidic property. It is an acylating agent, which converts thermally unstable compounds to its more volatile derivative and then made suitable for GC analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Heptafluorobutyryl)imidazole is used in gas chromatography for the determination of various pharmaceutical compounds like retronecine in biological matrixes. It acts as a reagent for derivatize amine-groups. It plays an essential role to reduce GC column degradation due to its non-acidic property. It is an acylating agent, which converts thermally unstable compounds to its more volatile derivative and then made suitable for GC analysis.

Solubility
Miscible with chloroform and methanol.

Notes
Hygroscopic and air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text