Thermo Scientific Chemicals

Tetraphenylethylene, 98%, Thermo Scientific Chemicals

Catalog number: L00222.06
5 g, Each
Thermo Scientific Chemicals

Tetraphenylethylene, 98%, Thermo Scientific Chemicals

Catalog number: L00222.06
5 g, Each
Quantity
Catalog number: L00222.06
also known as L00222-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
632-51-9
IUPAC Name
(1,2,2-triphenylethenyl)benzene
Molecular Formula
C26H20
InChI Key
JLZUZNKTTIRERF-UHFFFAOYSA-N
SMILES
C1=CC=C(C=C1)C(=C(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to cream
Assay (HPLC)
≥97.5%
Melting Point (clear melt)
222-228?C
Form
Crystalline powder
Identification (FTIR)
Conforms

Description

Tetraphenylethylene is used in construction and in the manufacture of medical equipment, packaging, and electrical appliances. And it is used to produce other chemical like 1,1,2,2-Tetraphenyl-ethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraphenylethylene is used in construction and in the manufacture of medical equipment, packaging, and electrical appliances. And it is used to produce other chemical like 1,1,2,2-Tetraphenyl-ethane.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text