Thermo Scientific Chemicals

Butadiene monoxide, 98%, Thermo Scientific Chemicals

Catalog number: L00238.14
25 g, Each
Thermo Scientific Chemicals

Butadiene monoxide, 98%, Thermo Scientific Chemicals

Catalog number: L00238.14
25 g, Each
Quantity
Catalog number: L00238.14
also known as L00238-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
930-22-3
IUPAC Name
2-ethenyloxirane
Molecular Formula
C4H6O
InChI Key
GXBYFVGCMPJVJX-UHFFFAOYNA-N
SMILES
C=CC1CO1
Refractive Index
1.4155-1.4200 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Form
Liquid

Description

Butadiene monoxide is used as key starting material for the synthesis of (-)-bulgecinine and (+)-broussonetine G.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butadiene monoxide is used as key starting material for the synthesis of (-)-bulgecinine and (+)-broussonetine G.

Solubility
Miscible with ethanol, ethyl ether, benzene and organic solvents.

Notes
Moisture sensitive. Store in cool place. Incompatible with acids, bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text