Thermo Scientific Chemicals

tert-Butyl carbamate, 98+%, Thermo Scientific Chemicals

Catalog number: L00259.06
5 g, Each
Thermo Scientific Chemicals

tert-Butyl carbamate, 98+%, Thermo Scientific Chemicals

Catalog number: L00259.06
5 g, Each
Quantity
Catalog number: L00259.06
also known as L00259-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4248-19-5
IUPAC Name
tert-butyl carbamate
Molecular Formula
C5H11NO2
InChI Key
LFKDJXLFVYVEFG-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(N)=O
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale yellow or pale pink
Assay (GC)
≥98.0%
Melting Point (clear melt)
105-110?C

Description

tert-Butyl carbamate is used in palladium-catalyzed synthesis of N-Boc-protected anilines. It was used in the synthesis of tetrasubstituted pyrroles, functionalized with ester or ketone groups at C-3 position.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl carbamate is used in palladium-catalyzed synthesis of N-Boc-protected anilines. It was used in the synthesis of tetrasubstituted pyrroles, functionalized with ester or ketone groups at C-3 position.

Solubility
Soluble in methylene chloride, chloroform, and alcohols. Slightly soluble in petroleum ether and water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text