Thermo Scientific Chemicals

Phenyl formate, 95%, Thermo Scientific Chemicals

Catalog number: L00289.06
5 g, Each
Thermo Scientific Chemicals

Phenyl formate, 95%, Thermo Scientific Chemicals

Catalog number: L00289.06
5 g, Each
Quantity
Catalog number: L00289.06
also known as L00289-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1864-94-4
IUPAC Name
phenyl formate
Molecular Formula
C7H6O2
InChI Key
GEOWCLRLLWTHDN-UHFFFAOYSA-N
SMILES
O=COC1=CC=CC=C1
Refractive Index
1.5060-1.5150 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥94.0%
Identification (FTIR)
Conforms
Form
Liquid

Description

Phenyl formate is widely used in the palladium-catalyzed carbonylation of aryl, alkenyl and allyl halides. It is also used as a reagent for the formylation of amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl formate is widely used in the palladium-catalyzed carbonylation of aryl, alkenyl and allyl halides. It is also used as a reagent for the formylation of amines.

Solubility
Immiscible with water, alcohol, ether and benzene.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text