Thermo Scientific Chemicals

1,8-Bis(dimethylamino)naphthalene, 98+%, Thermo Scientific Chemicals

Catalog number: L00313.09
10 g, Each
Thermo Scientific Chemicals

1,8-Bis(dimethylamino)naphthalene, 98+%, Thermo Scientific Chemicals

Catalog number: L00313.09
10 g, Each
Quantity
Catalog number: L00313.09
also known as L00313-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20734-58-1
IUPAC Name
N1,N1,N8,N8-tetramethylnaphthalene-1,8-diamine
Molecular Formula
C14H18N2
InChI Key
GJFNRSDCSTVPCJ-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=CC2=CC=CC(N(C)C)=C12
Form
Crystalline powder
Melting Point (clear melt)
46-52?C
Appearance (Color)
White to cream to yellow to pink or pink-brown
Assay (GC)
≥98.0%

Description

1,8-Bis(dimethylamino)naphthalene is used in the synthesis of C-substituted t-BuNH-8,9-R,R?-nido-7,8,9-C3B8H9 (R,R? = H,H; MeH; Me,Me; Ph,H and Ph,Ph) tricarbollide compounds. It was also used in the preparation of saturated fluoroalkyl(hydrido) complexes of Iridium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Bis(dimethylamino)naphthalene is used in the synthesis of C-substituted t-BuNH-8,9-R,R′-nido-7,8,9-C3B8H9 (R,R′ = H,H; MeH; Me,Me; Ph,H and Ph,Ph) tricarbollide compounds. It was also used in the preparation of saturated fluoroalkyl(hydrido) complexes of Iridium.

Solubility
Soluble in organic solvents.

Notes
Air & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text