Thermo Scientific Chemicals

4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals

Catalog number: L00314.18
50 g, Each
Thermo Scientific Chemicals

4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals

Catalog number: L00314.18
50 g, Each
Quantity
Catalog number: L00314.18
also known as L00314-18
Price (TWD)

Chemical Identifiers

CAS
830-03-5
IUPAC Name
4-nitrophenyl acetate
Molecular Formula
C8H7NO4
InChI Key
QAUUDNIGJSLPSX-UHFFFAOYSA-N
SMILES
CC(=O)OC1=CC=C(C=C1)[N+]([O-])=O
Appearance (Color)
White to pale cream to cream to yellow to pale brown
Form
Crystals or powder or crystalline powder or fused solid
Identification (FTIR)
Conforms
Assay (GC)
≥96.0%

Description

4-Nitrophenyl acetate is used with iodoacetic acid for reductive cleavage of methionine-containing peptides. It is also used as a substrate that has been used in assays for esterase and lipase activity. Inorganic complexes have been evaluated for their methanolysis or hydrolysis activity using p-nitrophenyl acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl acetate is used with iodoacetic acid for reductive cleavage of methionine-containing peptides. It is also used as a substrate that has been used in assays for esterase and lipase activity. Inorganic complexes have been evaluated for their methanolysis or hydrolysis activity using p-nitrophenyl acetate.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text