Thermo Scientific Chemicals

4-Nitrophenyl formate, 98%, Thermo Scientific Chemicals

Catalog number: L00316.14
25 g, Each
Thermo Scientific Chemicals

4-Nitrophenyl formate, 98%, Thermo Scientific Chemicals

Catalog number: L00316.14
25 g, Each
Quantity
Catalog number: L00316.14
also known as L00316-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1865-01-6
IUPAC Name
4-nitrophenyl formate
Molecular Formula
C7H5NO4
InChI Key
IEXRKQFZXJSHOB-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(OC=O)C=C1
Appearance (Color)
White to cream to pale yellow
Assay (HPLC)
≥97.5%
Form
Crystals or powder or crystalline powder or fused solid

Description

Convenient and selective reagent for the selective formylation of terminal amino groups in such basic amino acids as ornithine or lysine and formylation of α-amino groups in peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Convenient and selective reagent for the selective formylation of terminal amino groups in such basic amino acids as ornithine or lysine and formylation of α-amino groups in peptides.

Solubility
Hydrolyzes in water. Soluble in dichloromethane.

Notes
Moisture and light sensitive. Store away from water/moisture, oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark. Store under inert gas. Protect from exposure to light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text