Thermo Scientific Chemicals

N-Ethylmaleimide, 98+%, Thermo Scientific Chemicals

Catalog number: L00355.09
10 g, Each
Thermo Scientific Chemicals

N-Ethylmaleimide, 98+%, Thermo Scientific Chemicals

Catalog number: L00355.09
10 g, Each
Quantity
Catalog number: L00355.09
also known as L00355-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
128-53-0
IUPAC Name
1-ethyl-2,5-dihydro-1H-pyrrole-2,5-dione
Molecular Formula
C6H7NO2
InChI Key
HDFGOPSGAURCEO-UHFFFAOYSA-N
SMILES
CCN1C(=O)C=CC1=O
Identification (FTIR)
Conforms (UK-sourced material)
Melting Point (clear melt)
40.0-46.0°C
Appearance (Color)
White to cream or pale yellow
Form
Crystals or powder or crystalline powder and/or lumps/chunks or fused solid
Assay (GC)
≥98.0% (UK-sourced material)

Description

Cysteine alkylating agent N-Ethylmaleimide is a protein thiol modifier which inhibits apoptotic DNA fragmentation. It is a sulfhydryl reagent and finds application in experimental biochemical studies as well as in enzymology. It is involved in the modification of cysteine residues in proteins and peptides. It acts as a Michael acceptor and reacts with nucelophiles like thiols. It is employed in the inhibition of Mg2+-dependent internucleosomal DNA fragmentation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cysteine alkylating agent N-Ethylmaleimide is a protein thiol modifier which inhibits apoptotic DNA fragmentation. It is a sulfhydryl reagent and finds application in experimental biochemical studies as well as in enzymology. It is involved in the modification of cysteine residues in proteins and peptides. It acts as a Michael acceptor and reacts with nucelophiles like thiols. It is employed in the inhibition of Mg2+-dependent internucleosomal DNA fragmentation.

Solubility
Soluble in ethanol, water, chloroform, methanol and toluene.

Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text