N,N-Dimethyl-3-nitroaniline, 98%, Thermo Scientific Chemicals
N,N-Dimethyl-3-nitroaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Dimethyl-3-nitroaniline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L00401.06
also known as L00401-06
Price (USD)/ Each
41.30
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
41.30
Add to cart
N,N-Dimethyl-3-nitroaniline, 98%, Thermo Scientific Chemicals
Catalog numberL00401.06
Price (USD)/ Each
41.30
-
Add to cart
Chemical Identifiers
CAS619-31-8
IUPAC NameN,N-dimethyl-3-nitroaniline
Molecular FormulaC8H10N2O2
InChI KeyCJDICMLSLYHRPT-UHFFFAOYSA-N
SMILESCN(C)C1=CC=CC(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)56.0-62.0?C
Appearance (Color)Orange to red
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only