Thermo Scientific Chemicals

N,N-Dimethyl-m-phenylenediamine dihydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: L00403.06
5 g, Each
Thermo Scientific Chemicals

N,N-Dimethyl-m-phenylenediamine dihydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: L00403.06
5 g, Each
Quantity
Catalog number: L00403.06
also known as L00403-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3575-32-4
IUPAC Name
N1,N1-dimethylbenzene-1,3-diamine dihydrochloride
Molecular Formula
C8H14Cl2N2
InChI Key
BZJPIQKDEGXVFG-UHFFFAOYSA-N
SMILES
Cl.Cl.CN(C)C1=CC=CC(N)=C1
Form
Crystals or powder or crystalline powder or lumps or fused solid
Appearance (Color)
White to cream to yellow to pale brown or blue to pink to pale gray to gray
Assay (Titration ex Chloride)
≥98.5 to ≤101.5%

Description

it can be used to produce N-(4,5,6-trimethoxy-2,4-dimethyl-tetrahydro-pyran-3-yl)-formamide with 4,5,6-trimethoxy-2,4-dimethyl-tetrahydro-pyran-3-ylamine. This reaction will need solvent tetrahydrofuran. it is applied as a selective formylating agent for one of two amino groups in such basic amino acids as ornithine or lysine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
it can be used to produce N-(4,5,6-trimethoxy-2,4-dimethyl-tetrahydro-pyran-3-yl)-formamide with 4,5,6-trimethoxy-2,4-dimethyl-tetrahydro-pyran-3-ylamine. This reaction will need solvent tetrahydrofuran. it is applied as a selective formylating agent for one of two amino groups in such basic amino acids as ornithine or lysine.

Solubility
Soluble in water, chloroform and dichloromethane. Slightly soluble in ethyl acetate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text