Thermo Scientific Chemicals

Hexaphenylbenzene, 98+%, Thermo Scientific Chemicals

Catalog number: L00446.06
5 g, Each
Thermo Scientific Chemicals

Hexaphenylbenzene, 98+%, Thermo Scientific Chemicals

Catalog number: L00446.06
5 g, Each
Quantity
Catalog number: L00446.06
also known as L00446-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
992-04-1
IUPAC Name
2,3,4,5,6-pentaphenyl-1,1'-biphenyl
Molecular Formula
C42H30
InChI Key
QBHWPVJPWQGYDS-UHFFFAOYSA-N
SMILES
C1=CC=C(C=C1)C1=C(C2=CC=CC=C2)C(C2=CC=CC=C2)=C(C2=CC=CC=C2)C(C2=CC=CC=C2)=C1C1=CC=CC=C1
Appearance (Color)
White to pale cream to cream to brown
Purity (DSC)
≥98.0%
Melting Point (DSC)
450.0-455.0?C
Form
Crystals or powder or crystalline powder

Description

Hexaphenylbenzene was used to prepare the fluorescent nanorods used for the detection of trinitrotoulene (TNT).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexaphenylbenzene was used to prepare the fluorescent nanorods used for the detection of trinitrotoulene (TNT).

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text