Thermo Scientific Chemicals

2,4,6-Trimethylpyrylium tetrafluoroborate, 98+%, Thermo Scientific Chemicals

Catalog number: L00467.06
5 g, Each
Thermo Scientific Chemicals

2,4,6-Trimethylpyrylium tetrafluoroborate, 98+%, Thermo Scientific Chemicals

Catalog number: L00467.06
5 g, Each
Quantity
Catalog number: L00467.06
also known as L00467-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
773-01-3
IUPAC Name
2,4,6-trimethyl-1λ⁴-pyran-1-ylium; tetrafluoroboranuide
Molecular Formula
C8H11BF4O
InChI Key
PYOPRIFMELCENN-UHFFFAOYSA-N
SMILES
F[B-](F)(F)F.CC1=CC(C)=[O+]C(C)=C1
Appearance (Color)
White to cream to pale green
Form
Crystals or powder or crystalline powder or fused solid
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Melting Point (clear melt)
220.0-228.0?C

Description

It is used in the radical Arylation Reactions of 4, 6, 8-Trimethylazulene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the radical Arylation Reactions of 4, 6, 8-Trimethylazulene.

Solubility
Soluble in water and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text