It is used in the radical Arylation Reactions of 4, 6, 8-Trimethylazulene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the radical Arylation Reactions of 4, 6, 8-Trimethylazulene.
Solubility
Soluble in water and methanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
P. Mäding, et al. No-carrier-added 11C-labelling of benzenoid compounds in ring positions by condensation of nitro-[11C]methane with pyrylium salts.J. Labelled Compd. Radiopharm.,2000,43,(6), 565-583.
Anne Andrée Sophie Briquet, et al. Radical Arylation Reactions of 4, 6, 8-Trimethylazulene.Helvetica Chimica Acta.,1994,77,(6), 1577-1584.