4-Methoxybenzhydrol, 98+%, Thermo Scientific Chemicals
4-Methoxybenzhydrol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methoxybenzhydrol, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L00468.06
also known as L00468-06
Price (USD)/ Each
43.80
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
43.80
Add to cart
4-Methoxybenzhydrol, 98+%, Thermo Scientific Chemicals
Catalog numberL00468.06
Price (USD)/ Each
43.80
-
Add to cart
Chemical Identifiers
CAS720-44-5
IUPAC Name(4-methoxyphenyl)(phenyl)methanol
Molecular FormulaC14H14O2
InChI KeyBEGZWXVLBIZFKQ-UHFFFAOYNA-N
SMILESCOC1=CC=C(C=C1)C(O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to red
FormCrystals or powder or crystalline powder or lumps or fused solid
Assay (GC)≥98.0%
Melting Point (clear melt)63.0-69.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only