Platinum, 10% on carbon, dry, Thermo Scientific Chemicals
Platinum, 10% on carbon, dry, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum, 10% on carbon, dry, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L00469.03
also known as L00469-03
Price (USD)/ Each
65.50
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
65.50
Add to cart
Platinum, 10% on carbon, dry, Thermo Scientific Chemicals
Catalog numberL00469.03
Price (USD)/ Each
65.50
-
Add to cart
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Formpowder
Assay (unspecified)Pt 9-11%
Loss on Drying≤ 5.0%
CommentSupport is type 18 carbon

Platinum, 10% on carbon, is used as a catalyst for the hydrogenation of alkenes, alkynes, nitro groups, ketones and aromatics. It is also used for hydrogenolysis of cyclopropanes. It is involved in the catalyzes oxidation of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Platinum, 10% on carbon, is used as a catalyst for the hydrogenation of alkenes, alkynes, nitro groups, ketones and aromatics. It is also used for hydrogenolysis of cyclopropanes. It is involved in the catalyzes oxidation of alcohols.

Solubility
Insoluble in water.

Notes
Store in a cool location. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Chen, S.; Qi, P.; Chen, J.; Yuan, Y. Platinum nanoparticles supported on N-doped carbon nanotubes for the selective oxidation of glycerol to glyceric acid in a base-free aqueous solution. RSC Adv. 2015, 5, 31566-31574.
  2. Sharma, R.; Kar, K. K. Particle size and crystallographic orientation controlled electrodeposition of platinum nanoparticles on carbon nanotubes. Electrochim. Acta 2015, 156, 199-206.