Thermo Scientific Chemicals

2,2-Dimethyl-1,3-dioxolane, 98+%, Thermo Scientific Chemicals

Catalog number: L00482.06
5 g, Each
Thermo Scientific Chemicals

2,2-Dimethyl-1,3-dioxolane, 98+%, Thermo Scientific Chemicals

Catalog number: L00482.06
5 g, Each
Quantity
Catalog number: L00482.06
also known as L00482-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2916-31-6
IUPAC Name
2,2-dimethyl-1,3-dioxolane
Molecular Formula
C5H10O2
InChI Key
SIJBDWPVNAYVGY-UHFFFAOYSA-N
SMILES
CC1(C)OCCO1
Appearance (Color)
Clear colorless
Form
Liquid
Refractive Index
1.3965-1.4005 @ 20?C
Assay (GC)
≥98.0%

Description

2,2-Dimethyl-1,3-dioxolane is used as organic intermediates, fine chemicals, pharmaceutical research and development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Dimethyl-1,3-dioxolane is used as organic intermediates, fine chemicals, pharmaceutical research and development.

Solubility
Insoluble in water.

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text