Thermo Scientific Chemicals

Tetraphenylphosphine imide, 97%, Thermo Scientific Chemicals

Catalog number: L00496.03
1 g, Each
Thermo Scientific Chemicals

Tetraphenylphosphine imide, 97%, Thermo Scientific Chemicals

Catalog number: L00496.03
1 g, Each
Quantity
Catalog number: L00496.03
also known as L00496-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2325-27-1
IUPAC Name
N,1,1,1-tetraphenyl-λ⁵-phosphanimine
Molecular Formula
C24H20NP
InChI Key
PTLOPIHJOPWUNN-UHFFFAOYSA-N
SMILES
C1=CC=C(C=C1)N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Purity (DSC)
≥96.0%
Form
Crystals or powder or crystalline powder or fused solid
Appearance (Color)
Pale yellow to yellow to green
Assay (GC)
≥96.0%

Description

Has been used as a protected form of aniline for the preparation of monoalkylanilines by alkylation and hydrolysis and as an intermediate for the preparation of isocyanates and isothiocyanates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Has been used as a protected form of aniline for the preparation of monoalkylanilines by alkylation and hydrolysis and as an intermediate for the preparation of isocyanates and isothiocyanates.

Solubility
Insoluble in water. Soluble in chloroform 25 mg/mL.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text