Thermo Scientific Chemicals

1-Boc-imidazole, 98%, Thermo Scientific Chemicals

Catalog number: L00506.14
25 g, Each
Thermo Scientific Chemicals

1-Boc-imidazole, 98%, Thermo Scientific Chemicals

Catalog number: L00506.14
25 g, Each
Quantity
Catalog number: L00506.14
also known as L00506-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
49761-82-2
IUPAC Name
tert-butyl 1H-imidazole-1-carboxylate
Molecular Formula
C8H12N2O2
InChI Key
MTBKGWHHOBJMHJ-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)N1C=CN=C1
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Appearance (Color)
White
Melting Point (clear melt)
42.5-48.5?C

Description

1-Boc-imidazole is used in the preparation of water soluble polyrotaxane, via modification of α-cyclodextrin in the presence of KOH and N,N?-carbonyldiimidazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-imidazole is used in the preparation of water soluble polyrotaxane, via modification of α-cyclodextrin in the presence of KOH and N,N′-carbonyldiimidazole.

Solubility
Soluble in hexane, acetone, and DMF.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text