Thermo Scientific Chemicals

Methyldiphenylsulfonium tetrafluoroborate, 95%, Thermo Scientific Chemicals

Catalog number: L00512.14
25 g, Each
Thermo Scientific Chemicals

Methyldiphenylsulfonium tetrafluoroborate, 95%, Thermo Scientific Chemicals

Catalog number: L00512.14
25 g, Each
Quantity
Catalog number: L00512.14
also known as L00512-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10504-60-6
IUPAC Name
methyldiphenylsulfanium; tetrafluoroboranuide
Molecular Formula
C13H13BF4S
InChI Key
LHAMVDBAOJCBDW-UHFFFAOYSA-N
SMILES
F[B-](F)(F)F.C[S+](C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White
Assay (HPLC)
≥94.0%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
60-68?C

Description

Methyldiphenylsulfonium tetrafluoroborate is a Methylene transfer agent. It is also used in alkyl diphenylsulfonium salts in place of trialkyl avoids sulfur-containing by-products arising from the Sommelet-Hauser rearrangement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyldiphenylsulfonium tetrafluoroborate is a Methylene transfer agent. It is also used in alkyl diphenylsulfonium salts in place of trialkyl avoids sulfur-containing by-products arising from the Sommelet-Hauser rearrangement.

Solubility
Soluble in water, alcohol.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text