Thermo Scientific Chemicals

Chlorodimethyl(pentafluorophenyl)silane, 96%, Thermo Scientific Chemicals

Catalog number: L00550.03
1 g, Each
Thermo Scientific Chemicals

Chlorodimethyl(pentafluorophenyl)silane, 96%, Thermo Scientific Chemicals

Catalog number: L00550.03
1 g, Each
Quantity
Catalog number: L00550.03
also known as L00550-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20082-71-7
IUPAC Name
chlorodimethyl(2,3,4,5,6-pentafluorophenyl)silane
Molecular Formula
C8H6ClF5Si
InChI Key
PQRFRTCWNCVQHI-UHFFFAOYSA-N
SMILES
C[Si](C)(Cl)C1=C(F)C(F)=C(F)C(F)=C1F
Assay (GC)
≥95.0%
Form
Liquid
Appearance (Color)
Clear colorless
Refractive Index
1.4435-1.4495 @ 20?C

Description

It is used as a GC reagent for detection of sterols at low concentration as flophemesyl derivatives, readily detected by electron capture. Chlorodimethyl(pentafluorophenyl)silane is an extremely sensitive derivatizing agent for the analysis of sterols and lower aliphatic alcohols by electron capture gas chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a GC reagent for detection of sterols at low concentration as flophemesyl derivatives, readily detected by electron capture. Chlorodimethyl(pentafluorophenyl)silane is an extremely sensitive derivatizing agent for the analysis of sterols and lower aliphatic alcohols by electron capture gas chromatography.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from strong bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text