Thermo Scientific Chemicals

N,N,N',N'-Tetramethyl-p-phenylenediamine, 98+%, Thermo Scientific Chemicals

Catalog number: L00559.09
10 g, Each
Thermo Scientific Chemicals

N,N,N',N'-Tetramethyl-p-phenylenediamine, 98+%, Thermo Scientific Chemicals

Catalog number: L00559.09
10 g, Each
Quantity
Catalog number: L00559.09
also known as L00559-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-22-1
IUPAC Name
N1,N1,N4,N4-tetramethylbenzene-1,4-diamine
Molecular Formula
C10H16N2
InChI Key
CJAOGUFAAWZWNI-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=C(C=C1)N(C)C
Identification (FTIR)
Conforms
Melting Point (clear melt)
47.0-54.0?C
Assay (GC)
≥98.0%
Appearance (Color)
White to cream to red to dark brown to brown or pale blue to blue to purple to pale gray to gray
Form
Crystals or powder or crystalline powder or lumps or fused solid or flakes or granules

Description

N,N,N',N'-Tetramethyl-p-phenylenediamine is a reducing co-substrate for heme peroxidases. It is used for detection of peroxidases on polyacrylamide gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N,N′,N′-Tetramethyl-p-phenylenediamine is a reducing co-substrate for heme peroxidases. It is used for detection of peroxidases on polyacrylamide gels.

Solubility
Slightly soluble in cold, more in hot water.

Notes
Air & Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text