Thermo Scientific Chemicals

Diethyl benzylidenemalonate, 98%, Thermo Scientific Chemicals

Catalog number: L00561.22
100 g, Each
Thermo Scientific Chemicals

Diethyl benzylidenemalonate, 98%, Thermo Scientific Chemicals

Catalog number: L00561.22
100 g, Each
Quantity
Catalog number: L00561.22
also known as L00561-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5292-53-5
IUPAC Name
1,3-diethyl 2-(phenylmethylidene)propanedioate
Molecular Formula
C14H16O4
InChI Key
VUWPIBNKJSEYIN-UHFFFAOYSA-N
SMILES
CCOC(=O)C(=CC1=CC=CC=C1)C(=O)OCC
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.5340-1.5380 @ 20°C

Description

Diethyl benzylidenemalonate is used as a pharmaceutical intermediate. It is also involved in the Conjugate addition of cyanide ion to arylidenemalonic esters provides a useful route to arylsuccinic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl benzylidenemalonate is used as a pharmaceutical intermediate. It is also involved in the Conjugate addition of cyanide ion to arylidenemalonic esters provides a useful route to arylsuccinic acids.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text