Thermo Scientific Chemicals

1,4-Anthraquinone, 94%, Thermo Scientific Chemicals

Catalog number: L00573.14
25 g, Each
Thermo Scientific Chemicals

1,4-Anthraquinone, 94%, Thermo Scientific Chemicals

Catalog number: L00573.14
25 g, Each
Quantity
Catalog number: L00573.14
also known as L00573-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
635-12-1
IUPAC Name
1,4-dihydroanthracene-1,4-dione
Molecular Formula
C14H8O2
InChI Key
LSOTZYUVGZKSHR-UHFFFAOYSA-N
SMILES
O=C1C=CC(=O)C2=C1C=C1C=CC=CC1=C2
Appearance (Color)
Orange to brown
Form
Powder
Assay (HPLC)
≥92.0%

Description

1,4-Anthraquinone is used in the preparation of 1-methyl-1,4,4a,12a-tetrahydronaphthacene-5,12-dione.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Anthraquinone is used in the preparation of 1-methyl-1,4,4a,12a-tetrahydronaphthacene-5,12-dione.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text