Thermo Scientific Chemicals

S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine, 98%, Thermo Scientific Chemicals

Catalog number: L00577.14
25 g, Each
Thermo Scientific Chemicals

S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine, 98%, Thermo Scientific Chemicals

Catalog number: L00577.14
25 g, Each
Quantity
Catalog number: L00577.14
also known as L00577-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22236-45-9
IUPAC Name
dimethyl[(4-methylbenzenesulfonyl)imino]-λ⁶-sulfanone
Molecular Formula
C9H13NO3S2
InChI Key
IRNAWARRPQUZDU-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)S(=O)(=O)N=S(C)(C)=O
Assay (HPLC)
≥97.5%
Form
Crystals or powder or crystalline powder
Appearance (Color)
White
Melting Point
166-172°C

Description

S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine, is used as methylene-transfer agent. The anion adds to carbonyl compounds to give epoxides which can react with a further mole of reagent with ring expansion to give 2,2-disubstituted oxetanes. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine, is used as methylene-transfer agent. The anion adds to carbonyl compounds to give epoxides which can react with a further mole of reagent with ring expansion to give 2,2-disubstituted oxetanes. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in EtOH, THF, DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text