Thermo Scientific Chemicals

Ethyl o-toluate, 98+%, Thermo Scientific Chemicals

Catalog number: L00598.14
25 g, Each
Thermo Scientific Chemicals

Ethyl o-toluate, 98+%, Thermo Scientific Chemicals

Catalog number: L00598.14
25 g, Each
Quantity
Catalog number: L00598.14
also known as L00598-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
87-24-1
IUPAC Name
ethyl 2-methylbenzoate
Molecular Formula
C10H12O2
InChI Key
SOUAXOGPALPTTC-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC=CC=C1C
Appearance (Color)
Clear colorless to pale green
Assay (GC)
≥98.0%
Refractive Index
1.5055-1.5095 @ 20?C
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text