Thermo Scientific Chemicals

Ethyl phenylpropiolate, 98+%, Thermo Scientific Chemicals

Catalog number: L00603.06
5 g, Each
Thermo Scientific Chemicals

Ethyl phenylpropiolate, 98+%, Thermo Scientific Chemicals

Catalog number: L00603.06
5 g, Each
Quantity
Catalog number: L00603.06
also known as L00603-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2216-94-6
IUPAC Name
ethyl 3-phenylprop-2-ynoate
Molecular Formula
C11H10O2
InChI Key
ACJOYTKWHPEIHW-UHFFFAOYSA-N
SMILES
CCOC(=O)C#CC1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.5500-1.5540 @ 20?C
Form
Liquid
Assay (GC)
>98.0%

Description

Ethyl phenylpropiolate, is an important organic intermediate. It is also used as an intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl phenylpropiolate, is an important organic intermediate. It is also used as an intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Heat sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text