Thermo Scientific Chemicals

N,N-Dimethylformamide di-n-propyl acetal, 97%, Thermo Scientific Chemicals

Catalog number: L00643.18
50 g, Each
Thermo Scientific Chemicals

N,N-Dimethylformamide di-n-propyl acetal, 97%, Thermo Scientific Chemicals

Catalog number: L00643.18
50 g, Each
Quantity
Catalog number: L00643.18
also known as L00643-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6006-65-1
Form
Liquid
Refractive Index
1.4070-1.4120 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%

Description

N,N-Dimethylformamide di-n-propyl acetal, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylformamide di-n-propyl acetal, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
It hydrolyzes with water.

Notes
Moisture, water Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text