Trimethylsulfonium tetrafluoroborate, is used as Methylene transfer agent and as a pharmaceutical intermediate. It is also used as a raw material in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Trimethylsulfonium tetrafluoroborate, is used as Methylene transfer agent and as a pharmaceutical intermediate. It is also used as a raw material in organic synthesis.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only
JO Valderrama.; RE Rojas. Mass connectivity index, a new molecular parameter for the estimation of ionic liquid properties. Fluid Phase Equilibria. 2010297,(1) , 107-112.
F Mahuteau-Betzer.; L Ghosez. Asymmetric [2+ 2+ 1] cyclopentannulation of olefins. Ring expansion of 2-N-methyl-N-tosyl-cyclobutanone. Tetrahedron. 200258,(35) , 6991-7000.
Methylene transfer agent, cf Trimethylsulfonium iodide, A12639.