Thermo Scientific Chemicals

Triethylsulfonium iodide, 97%, Thermo Scientific Chemicals

Catalog number: L00667.14
25 g, Each
Thermo Scientific Chemicals

Triethylsulfonium iodide, 97%, Thermo Scientific Chemicals

Catalog number: L00667.14
25 g, Each
Quantity
Catalog number: L00667.14
also known as L00667-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1829-92-1
IUPAC Name
triethylsulfanium iodide
Molecular Formula
C6H15IS
InChI Key
LKDQWVKWYGOVJW-UHFFFAOYSA-M
SMILES
[I-].CC[S+](CC)CC
Form
Crystals or powder or crystalline powder
Assay (Titration ex Iodide)
≥96.0 to ≤104.0%
Appearance (Color)
White to yellow or orange

Description

Triethylsulfonium iodide is used as an ethylidene-transfer reagent. It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylsulfonium iodide is used as an ethylidene-transfer reagent. It is used as a pharmaceutical intermediate.

Solubility
It is soluble in water.

Notes
Light, Heat Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text