Tetrabromocatechol, 96%, Thermo Scientific Chemicals
Tetrabromocatechol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrabromocatechol, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L00690.06
also known as L00690-06
Price (USD)/ Each
36.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
36.90
Add to cart
Tetrabromocatechol, 96%, Thermo Scientific Chemicals
Catalog numberL00690.06
Price (USD)/ Each
36.90
-
Add to cart
Chemical Identifiers
CAS488-47-1
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)Cream to brown
Assay (HPLC)≥95.0%
Tetrabromocatechol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrabromocatechol is used as a pharmaceutical intermediate.

Solubility
It is partly miscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong bases. Stable under recommended storage conditions.
RUO – Research Use Only

General References:

  1. A Panja. Unusual structural features in tetrabromocatechol-chelated dinuclear manganese(III) complex: Synthesis, electrochemistry and thermally induced valence tautomerism. Inorganic Chemistry Communications. 201224,(1) , 140-143.
  2. EW Lard.; RG Rice.; EE Stahly. Storage Stability of Unsaturated Polyesters. Ind. Eng. Chem. Prod. Res. Dev. 197110,(4) , 391-395.