Thermo Scientific Chemicals

Methyl cyclopentanecarboxylate, 97%, Thermo Scientific Chemicals

Catalog number: L00741.06
5 g, Each
Thermo Scientific Chemicals

Methyl cyclopentanecarboxylate, 97%, Thermo Scientific Chemicals

Catalog number: L00741.06
5 g, Each
Quantity
Catalog number: L00741.06
also known as L00741-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4630-80-2
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.4335-1.4385 @ 20?C
Identification (FTIR)
Conforms

Description

Methyl cyclopentanecarboxylate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl cyclopentanecarboxylate

Solubility
Difficult to mix.

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text