Thermo Scientific Chemicals

2,2-Difluorosuccinic acid, 94%, Thermo Scientific Chemicals

Catalog number: L00773.03
1 g, Each
Thermo Scientific Chemicals

2,2-Difluorosuccinic acid, 94%, Thermo Scientific Chemicals

Catalog number: L00773.03
1 g, Each
Quantity
Catalog number: L00773.03
also known as L00773-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
665-31-6
IUPAC Name
2,2-difluorobutanedioic acid
Molecular Formula
C4H4F2O4
InChI Key
ZYLFHISLYSHWRH-UHFFFAOYSA-N
SMILES
OC(=O)CC(F)(F)C(O)=O
Appearance (Color)
White to cream to brown
Assay (Aqueous acid-base Titration)
≥92.0 to ≤108.0%
Form
Crystals or powder or crystalline powder or granules
Melting Point (clear melt)
134.0-146.0?C
Assay (Silylated GC)
≥92.0%

Description

2,2-Difluorosuccinic acid is used in the preparation of fluorofumaric acid. It is also used in enolate alkylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluorosuccinic acid is used in the preparation of fluorofumaric acid. It is also used in enolate alkylation.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids, and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text